Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluorobenzamide 97%

Supplier: AMBEED, INC
Warning

A111892-100G A111892-10G A111892-25G A111892-500G
76765-888EA 31 USD InStock
76765-888 76765-890 76765-892 76765-894
2,6-Difluorobenzamide 97%
2,6-Difluorobenzamide
Precaución: for Research Use Only


Formula: C₇H₅F₂NO
MDL Number: MFCD00007972
CAS Number: 18063-03-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty