Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Methoxy-1H-indol-3-yl)methanamine 97%

Supplier: AMBEED, INC
Warning

A112092-100MG A112092-250MG
76765-914EA 36 USD InStock
76765-914 76765-916
(6-Methoxy-1H-indol-3-yl)methanamine 97%
(6-Methoxy-1H-indol-3-yl)methanamine
Precaución: for Research Use Only


Formula: C₁₀H₁₂N₂O
MDL Number: MFCD06657120
CAS Number: 887582-58-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty