Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trifluorobenzaldehyde 98% (by GC)

Supplier: AMBEED, INC
Warning

A112668-100G A112668-10G A112668-1G A112668-25G A112668-5G
76765-972EA 327 USD InStock
76765-972 76765-974 76765-976 76765-978 76765-980
2,4,6-Trifluorobenzaldehyde 98% (by GC)
2,4,6-Trifluorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₃F₃O
MDL Number: MFCD00061197
CAS Number: 58551-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty