Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-amino-2-bromobenzoate 98%

Supplier: AMBEED, INC
Danger

A114140-1G A114140-25G A114140-5G
76766-042EA 25 USD InStock
76766-042 76766-044 76766-046
Methyl 5-amino-2-bromobenzoate 98%
Methyl 5-amino-2-bromobenzoate
Precaución: for Research Use Only


Formula: C₈H₈BrNO₂
MDL Number: MFCD00085568
CAS Number: 6942-37-6
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty