Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1-methyl-1H-1,2,4-triazole 98%

Supplier: AMBEED, INC
Warning

A114429-1G A114429-250MG A114429-25G
76766-060EA 19 USD InStock
76766-060 76766-062 76766-064
5-Bromo-1-methyl-1H-1,2,4-triazole 98%
5-Bromo-1-methyl-1H-1,2,4-triazole
Precaución: For research use only.


Formula: C₃H₄BrN₃
MDL Number: MFCD11109756
CAS Number: 16681-72-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty