Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloronaphthalen-1-amine 96%

Supplier: AMBEED, INC
Warning

A116827-100MG A116827-1G A116827-250MG A116827-5G
76766-224EA 7 USD InStock
76766-224 76766-226 76766-228 77623-976
5-Chloronaphthalen-1-amine 96%
5-Chloronaphthalen-1-amine
Precaución: for Research Use Only


Formula: C10H8ClN
MDL Number: MFCD01026464
CAS Number: 2750-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty