Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(trifluoromethyl)benzonitrile 98%

Supplier: AMBEED, INC
Warning

A119199-100G A119199-1G A119199-25G A119199-5G
76766-340EA 163 USD InStock
76766-340 76766-342 76766-344 76766-346
3-Fluoro-5-(trifluoromethyl)benzonitrile 98%
3-Fluoro-5-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₃F₄N
MW: 189.11
Boiling Pt: 212
Melting Pt: 28
MDL Number: MFCD00061282
CAS Number: 149793-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty