Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3-chloro-2-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A119273-10G A119273-1G A119273-250MG A119273-5G
76766-362EA 359 USD InStock
76766-362 76766-364 76766-366 76766-368
6-Bromo-3-chloro-2-methylpyridine 98%
6-Bromo-3-chloro-2-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅BrClN
MDL Number: MFCD11110679
CAS Number: 944317-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty