Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-2-furaldehyde 98%

Supplier: AMBEED, INC
Warning

A119753-1000G A119753-100G A119753-25G A119753-500G
76766-398EA 117 USD InStock
76766-398 76766-400 76766-402 76766-404
5-Methyl-2-furaldehyde 98%
5-Methyl-2-furaldehyde
Precaución: for Research Use Only


Formula: C₆H₆O₂
MDL Number: MFCD00003232
CAS Number: 620-02-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty