Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-m-toluonitrile 97%

Supplier: AMBEED, INC
Warning

A120938-100G A120938-10G A120938-1G A120938-25G A120938-5G
76766-462EA 139 USD InStock
76766-462 76766-464 76766-466 76766-468 76766-470
6-Fluoro-m-toluonitrile 97%
6-Fluoro-m-toluonitrile
Precaución: for Research Use Only


Formula: C₈H₆FN
MW: 135.14
MDL Number: MFCD00134542
CAS Number: 64113-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty