Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-bromohexanoate 98%

Supplier: AMBEED, INC
Warning

A122718-100G A122718-10G A122718-25G A122718-500G A122718-5G
76766-622EA 59 USD InStock
76766-622 76766-624 76766-626 76766-628 76766-630
Ethyl-6-bromohexanoate 98%
Ethyl-6-bromohexanoate
Precaución: for Research Use Only


Formula: BrH₂C(CH₂)₄COOCH₂CH₃
MDL Number: MFCD00000270
CAS Number: 25542-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty