Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Phenylthiophene-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A122861-1G A122861-250MG A122861-5G
76766-658EA 44 USD InStock
76766-658 76766-660 76766-662
5-Phenylthiophene-2-carboxylic acid 97%
5-Phenylthiophene-2-carboxylic acid
Precaución: for Research Use Only


Formula: C11H8O2S
MW: 204.2450 g/mol
MDL Number: MFCD01313432
CAS Number: 19163-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty