Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-5-methylbenzonitrile 95%

Supplier: AMBEED, INC
Warning

A123089-1G A123089-250MG A123089-5G A123089-10G
76766-680EA 15 USD InStock
76766-680 76766-682 76766-684 77624-028
4-Bromo-2-fluoro-5-methylbenzonitrile 95%
4-Bromo-2-fluoro-5-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₈H₅BrFN
MW: 214.0300 g/mol
MDL Number: MFCD07782073
CAS Number: 916792-13-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty