Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-chloro-4-fluorophenol 95%

Supplier: AMBEED, INC
Warning

A123450-100MG A123450-10G A123450-1G A123450-250MG A123450-5G
76766-718EA 8 USD InStock
76766-718 76766-720 76766-722 76766-724 76766-726
5-Bromo-2-chloro-4-fluorophenol 95%
5-Bromo-2-chloro-4-fluorophenol
Precaución: for Research Use Only


Formula: C₆H₃BrClFO
MW: 225.4428 g/mol
MDL Number: MFCD08436979
CAS Number: 148254-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty