Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Thiophen-2-yl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A130278-1G A130278-250MG A130278-5G
76767-186EA 20 USD InStock
76767-186 76767-188 76767-190
4-(Thiophen-2-yl)benzoic acid 98%
4-(Thiophen-2-yl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₈O₂S
MDL Number: MFCD00690742
CAS Number: 29886-62-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty