Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nε-trifluoroacetyl-L-lysine 97%

Supplier: AMBEED, INC
Warning

A130353-10G A130353-5G A130353-25G A130353-500G
76767-204EA 6 USD InStock
76767-204 76767-206 77624-104 77624-106
Nε-trifluoroacetyl-L-lysine 97%
Nepsilon-trifluoroacetyl-L-lysine
Precaución: for Research Use Only


Formula: C₈H₁₃F₃N₂O₃
MW: 242.20 g/mol
MDL Number: MFCD00037223
CAS Number: 10009-20-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty