Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methylenanthic acid 95%

Supplier: AMBEED, INC
Danger

A131300-100MG A131300-1G A131300-250MG
76767-230EA 26 USD InStock
76767-230 76767-232 76767-234
6-Methylenanthic acid 95%
6-Methylenanthic acid
Precaución: for Research Use Only


Formula: C₈H₁₆O₂
MDL Number: MFCD00044083
CAS Number: 929-10-2
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty