Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,3-difluoroanisole 97%

Supplier: AMBEED, INC
Warning

A131388-100G A131388-10G A131388-1G A131388-25G A131388-5G
76767-238EA 125 USD InStock
76767-238 76767-240 76767-242 76767-244 76767-246
5-Bromo-2,3-difluoroanisole 97%
5-Bromo-2,3-difluoroanisole
Precaución: for Research Use Only


Formula: C₇H₅BrF₂O
MDL Number: MFCD01631352
CAS Number: 261762-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty