Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-fluorotoluene 98%

Supplier: AMBEED, INC
Warning

A133738-100G A133738-10G A133738-1G A133738-25G
76767-330EA 63 USD InStock
76767-330 76767-332 76767-334 76767-336 76767-338
3-Bromo-5-fluorotoluene 98%
3-Bromo-5-fluorotoluene


Formula: C₇H₆BrF
MDL Number: MFCD01861195
CAS Number: 202865-83-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty