Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3-nitro-4-quinolinol 97%

Supplier: AMBEED, INC
Warning

A133995-10G A133995-1G A133995-250MG A133995-5G
76767-342EA 33 USD InStock
76767-342 76767-344 76767-346 76767-348
6-Bromo-3-nitro-4-quinolinol 97%
6-Bromo-3-nitro-4-quinolinol
Precaución: for Research Use Only


Formula: C₉H₅BrN₂O₃
MW: 269.0516 g/mol
MDL Number: MFCD09800413
CAS Number: 853908-50-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty