Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A135003-1G A135003-250MG A135003-25G
76767-416EA 8 USD InStock
76767-416 76767-418 76767-420
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride 95%
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride


Formula: C₈H₇ClF₆N₂
MDL Number: MFCD03094165
CAS Number: 502496-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty