Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluorobenzonitrile 95%

Supplier: AMBEED, INC
Warning

A135737-100G A135737-10G A135737-25G A135737-500G A135737-5G
76767-456EA 88 USD InStock
76767-456 76767-458 76767-460 76767-462 76767-464
5-Chloro-2-fluorobenzonitrile 95%
5-Chloro-2-fluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃ClFN
MW: 155.56
MDL Number: MFCD01631434
CAS Number: 57381-34-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty