Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 5-amino-2-hydroxybenzoate 97%

Supplier: AMBEED, INC
Warning

A136079-1G A136079-5G
76767-510EA 177.18 USD InStock
76767-510 76767-512 76767-514 76767-516
Sodium 5-amino-2-hydroxybenzoate 97%
Sodium 5-amino-2-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₇H₆NNaO₃
MW: 175.12 g/mol
MDL Number: MFCD00070555
CAS Number: 35589-28-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty