Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-1H-benzo[d]imidazole 97%

Supplier: AMBEED, INC
Warning

A136803-10G A136803-1G A136803-250MG A136803-25G A136803-5G
76767-568EA 21 USD InStock
76767-568 76767-570 76767-572 76767-574 76767-576
6-Chloro-1H-benzo[d]imidazole 97%
5-Chlorobenzimidazole
Precaución: for Research Use Only


Formula: C₇H₅ClN₂
MDL Number: MFCD07432850
CAS Number: 4887-82-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty