Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-1-Boc-3-methylpiperazine 98%

Supplier: AMBEED, INC
Warning

A142121-100G A142121-10G A142121-1G A142121-25G A142121-500G A142121-5G
76767-832EA 69 USD InStock
76767-832 76767-834 76767-836 76767-838 76767-840 76767-842
(S)-(-)-1-Boc-3-methylpiperazine 98%
(S)-(-)-1-Boc-3-methylpiperazine
Precaución: for Research Use Only


Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD02683204
CAS Number: 147081-29-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty