Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-chlorobenzotrifluoride 98%

Supplier: AMBEED, INC
Warning

A142449-1000G A142449-100G A142449-10G A142449-25G A142449-500G A142449-5G
76767-858EA 165 USD InStock
76767-858 76767-860 76767-862 76767-864 76767-866 76767-868
5-Bromo-2-chlorobenzotrifluoride 98%
5-Bromo-2-chlorobenzotrifluoride
Precaución: for Research Use Only


Formula: C₇H₃BrClF₃
MW: 259.45
Boiling Pt: 198
Melting Pt: -1.4
Density: 1.7486
MDL Number: MFCD00000601
CAS Number: 445-01-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty