Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-5-methylbenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A147554-250MG A147554-1G
76768-030EA 41 USD InStock
76768-030 77624-308
4-Bromo-2-fluoro-5-methylbenzaldehyde 98%
4-Bromo-2-fluoro-5-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₆BrFO
MW: 217.04 g/mol
MDL Number: MFCD07782048
CAS Number: 916792-23-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty