Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropylbenzylamine 98%

Supplier: AMBEED, INC
Danger

A148136-100G A148136-10G A148136-1G A148136-250MG A148136-25G A148136-5G
76768-046EA 326 USD InStock
76768-046 76768-048 76768-050 76768-052 76768-054 76768-056
4-Isopropylbenzylamine 98%
4-Isopropylbenzylamine


Formula: C₁₀H₁₅N
MDL Number: MFCD00025580
CAS Number: 4395-73-7
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty