Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-hydroxypyridine 98%

Supplier: AMBEED, INC
Warning

A153122-10G A153122-1G A153122-250MG A153122-25G A153122-5G
76768-216EA 89 USD InStock
76768-216 76768-218 76768-220 76768-222 76768-224
2-Benzyloxy-5-hydroxypyridine 98%
2-Benzyloxy-5-hydroxypyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₂
MW: 201.2212 g/mol
MDL Number: MFCD06798281
CAS Number: 725256-57-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty