Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Piperidin-4yl)acetic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A155055-100G A155055-10G A155055-1G A155055-5G
76768-306EA 243 USD InStock
76768-306 76768-308 76768-310 76768-312
2-(Piperidin-4yl)acetic acid hydrochloride 95%
2-(Piperidin-4yl)acetic acid hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₄ClNO₂
MDL Number: MFCD03414187
CAS Number: 73415-84-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty