Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1-Piperidinylmethyl)phenol 97%

Supplier: AMBEED, INC
Warning

A156142-10G A156142-1G A156142-250MG A156142-25G A156142-5G
76768-338EA 48 USD InStock
76768-338 76768-340 76768-342 76768-344 77625-428
3-(1-Piperidinylmethyl)phenol 97%
3-(1-Piperidinylmethyl)phenol
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
MDL Number: MFCD00800250
CAS Number: 73279-04-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty