Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-chlorobenzothiazole 98%

Supplier: AMBEED, INC
Warning

A156412-100G A156412-10G A156412-500G
76768-358EA 33 USD InStock
76768-358 76768-360 76768-362
2-Amino-6-chlorobenzothiazole 98%
2-Amino-6-chlorobenzothiazole
Precaución: for Research Use Only


Formula: C₇H₅ClN₂S
MDL Number: MFCD00053557
CAS Number: 95-24-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty