Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-6-fluoro-1H-indole 98%

Supplier: AMBEED, INC
Warning

A157829-100MG A157829-1G A157829-250MG
76768-446EA 13 USD InStock
76768-446 76768-448 76768-450
5-Chloro-6-fluoro-1H-indole 98%
5-Chloro-6-fluoro-1H-indole
Precaución: for Research Use Only


Formula: C8H5ClFN
MW: 169.5800 g/mol
MDL Number: MFCD01074495
CAS Number: 169674-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty