Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Diacetylglycine anhydride 95%

Supplier: AMBEED, INC
Warning

A157857-10G A157857-1G A157857-250MG A157857-25G A157857-5G
76768-452EA 32 USD InStock
76768-452 76768-454 76768-456 76768-458 76768-460
N,N'-Diacetylglycine anhydride 95%
N,N'-Diacetylglycine anhydride
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₄
MW: 198.18 g/mol
MDL Number: MFCD00059789
CAS Number: 3027-05-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty