Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Miglitol 98%

Supplier: AMBEED, INC
Warning

A159516-1G A159516-1MG A159516-250MG
76768-506EA 28 USD InStock
76768-506 76768-508 76768-510
Miglitol 98%
Miglitol
Precaución: for Research Use Only


Formula: C₈H₁₇NO₅
MDL Number: MFCD00867240
CAS Number: 72432-03-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty