Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Acetylquinoline 98%

Supplier: AMBEED, INC
Warning

A162380-100MG A162380-1G A162380-250MG A162380-5G
76768-610EA 13 USD InStock
76768-610 76768-612 76768-614 76768-616
6-Acetylquinoline 98%
6-Acetylquinoline
Precaución: for Research Use Only


Formula: C₁₁H₉NO
MDL Number: MFCD00047614
CAS Number: 73013-68-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty