Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-5-fluoronicotinonitrile 98%

Supplier: AMBEED, INC
Warning

A162928-1000G A162928-100G A162928-10G A162928-25G A162928-500G A162928-5G
76768-652EA 397 USD InStock
76768-652 76768-654 76768-656 76768-658 76768-660 76768-662
2,6-Dichloro-5-fluoronicotinonitrile 98%
2,6-Dichloro-5-fluoronicotinonitrile
Precaución: for Research Use Only


Formula: C₆HCl₂FN₂
MW: 190.99 g/mol
MDL Number: MFCD01863289
CAS Number: 82671-02-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty