Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)hydrazine-1-carbothioamide 95%

Supplier: AMBEED, INC
Danger

A1636505-1G A1636505-250MG A1636505-5G
76768-722EA 77 USD InStock
76768-722 76768-724 76768-726
2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)hydrazine-1-carbothioamide 95%
2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)hydrazine-1-carbothioamide
Precaución: for Research Use Only


Formula: C₁₄H₂₀BN₃O₃S
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty