Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Glutamic acid 5-ethyl ester 97%

Supplier: AMBEED, INC
Warning

A165852-100G A165852-10G A165852-1G A165852-25G A165852-5G
76768-814EA 69 USD InStock
76768-814 76768-816 76768-818 76768-820 76768-822
L-Glutamic acid 5-ethyl ester 97%
L-Glutamic acid 5-ethyl ester
Precaución: for Research Use Only


Formula: C₇H₁₃NO₄
MDL Number: MFCD00037217
CAS Number: 1119-33-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty