Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)cyclohexanamine 95%

Supplier: AMBEED, INC
Warning

A1670706-1G A1670706-250MG A1670706-5G
76768-914EA 85 USD InStock
76768-914 76768-916 76768-918
N-(2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)cyclohexanamine 95%
N-(2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)cyclohexanamine
Precaución: for Research Use Only


Formula: C₁₉H₂₉BFNO₂
CAS Number: 2096340-26-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty