Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoic acid 95%

Supplier: AMBEED, INC
Warning

A1681591-100MG A1681591-1G A1681591-250MG A1681591-5G
76769-018EA 43 USD InStock
76769-018 76769-020 76769-022 76769-024
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoic acid 95%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoic acid
Precaución: for Research Use Only


Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty