Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzothioamide 97%

Supplier: AMBEED, INC
Warning

A1681609-1G A1681609-250MG A1681609-5G
76769-032EA 190 USD InStock
76769-032 76769-034 76769-036
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzothioamide 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzothioamide
Precaución: for Research Use Only


Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty