Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-(4,7-Dibromo-3-methoxynaphthalene-1,6-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) 95%

Supplier: AMBEED, INC
Warning

A1688713-100MG A1688713-1G A1688713-250MG
76769-156EA 43 USD InStock
76769-156 76769-158 76769-160
2,2'-(4,7-Dibromo-3-methoxynaphthalene-1,6-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) 95%
2,2'-(4,7-Dibromo-3-methoxynaphthalene-1,6-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
Precaución: for Research Use Only


Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty