Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-1H-indole-2,3-dicarboxylate 95%

Supplier: AMBEED, INC
Warning

A170496-100MG A170496-250MG A170496-1G
76769-204EA 16 USD InStock
76769-204 76769-206 77625-740
Dimethyl-1H-indole-2,3-dicarboxylate 95%
Dimethyl-1H-indole-2,3-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₄
MDL Number: MFCD00082731
CAS Number: 54781-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty