Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-iodo-m-xylene 98%

Supplier: AMBEED, INC
Warning

A176164-100G A176164-10G A176164-1G A176164-25G A176164-500G A176164-5G
76769-406EA 64 USD InStock
76769-406 76769-408 76769-410 76769-412 76769-414 76769-416
5-Bromo-2-iodo-m-xylene 98%
5-Bromo-2-iodo-m-xylene
Precaución: for Research Use Only


Formula: C₈H₈BrI
MW: 310.96
MDL Number: MFCD03094656
CAS Number: 206559-43-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty