Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Diaminophenol 97%

Supplier: AMBEED, INC
Warning

A178603-10G A178603-1G A178603-250MG A178603-25G A178603-5G
76769-484EA 114 USD InStock
76769-484 76769-486 76769-488 76769-490 76769-492
2,3-Diaminophenol 97%
2,3-Diaminophenol
Precaución: for Research Use Only


Formula: C₆H₈N₂O
MDL Number: MFCD00075199
CAS Number: 59649-56-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty