Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-chloro-2-methylpyridine 97%

Supplier: AMBEED, INC
Warning

A183200-1G A183200-250MG A183200-25G A183200-5G
76769-626EA 21 USD InStock
76769-626 76769-628 76769-630 76769-632
5-Bromo-3-chloro-2-methylpyridine 97%
5-Bromo-3-chloro-2-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅BrClN
MW: 206.48 g/mol
MDL Number: MFCD09031417
CAS Number: 914358-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty