Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-chlorobenzoylformate 97%

Supplier: AMBEED, INC
Warning

A183478-10G A183478-1G A183478-5G
76769-634EA 29 USD InStock
76769-634 76769-636 76769-638
Ethyl-4-chlorobenzoylformate 97%
Ethyl 4-chlorobenzoylformate
Precaución: for Research Use Only


Formula: C₁₀H₉ClO₃
MW: 212.6300 g/mol
MDL Number: MFCD01319615
CAS Number: 34966-48-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty