Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl-3-oxopyrrolidine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A184899-100G A184899-10G A184899-1G A184899-25G A184899-500G A184899-5G
76769-698EA 116 USD InStock
76769-698 76769-700 76769-702 76769-704 76769-706 76769-708
Benzyl-3-oxopyrrolidine-1-carboxylate 97%
Benzyl-3-oxopyrrolidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₃NO₃
MW: 219.2365 g/mol
MDL Number: MFCD03001711
CAS Number: 130312-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty