Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-(trifluoromethyl)benzonitrile 98%

Supplier: AMBEED, INC
Warning

A186260-100G A186260-10G A186260-1G A186260-25G A186260-5G
76769-746EA 183 USD InStock
76769-746 76769-748 76769-750 76769-752 76769-754
3-Chloro-5-(trifluoromethyl)benzonitrile 98%
3-Chloro-5-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₃ClF₃N
MDL Number: MFCD04115854
CAS Number: 693245-52-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty